Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eip93F
Entrez Gene ID
44936
Gene Name
Ecdysone-induced protein 93F
Gene Aliases
CG18389, Dmel\CG18389, Dmel_CG18389, E93, Eip93, Eip93f, anon-WO02059370.58, eip93F, eip93f, l(3)93, l(3)ry93, lincRNA.833, psg11
Chromosome Location
Chr.3R: 21949114 - 22014539 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264490
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104395.2 NP_001097865.1 6 - 7 1591 90
NM_001275888.1 NP_001262817.1 5 - 6 1437 90
NM_143745.3 NP_652002.2 5 - 6 1437 90
GenBank mRNA BT010034.1 - 6 - 7 1591 90
BT031093.1 - 4 - 5 1355 90

View Full Product Details