Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itpr
Entrez Gene ID
40664
Gene Name
Inositol 1,4,5,-trisphosphate receptor
Gene Aliases
CG1063, DIP, DmInsP[[3R]], DmInsP[[3]]R, Dmel\CG1063, Dmel_CG1063, IP, IP3R, IP[[3]] receptor, IP[[3]]R, ITP-r83A, ITPR1, Ins3PR, InsP(3), InsP3, InsP3R, InsP[[3]], InsP[[3]]R, Insp3R, Ip3R, Itp-R83A, Itp-r, Itp-r-83A, Itp-r42B, Itp-r83A, Itp-r[1], anon-WO02059370.49, dIP3R, dIP[[3]]R, dip, itpr, itpr-83, l(3)05616, l(3)itpr, l(3)j5B4
Chromosome Location
Chr.3R: 5517117 - 5539411 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010051
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300251.1 NP_001287180.1 8 - 9 3702 65
NM_169060.2 NP_730941.1 8 - 9 3657 65
NM_169061.2 NP_730942.1 8 - 9 3657 65

View Full Product Details