Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Spec2
- Entrez Gene ID
-
40685
- Gene Name
- CG14672-PA
- Gene Aliases
-
BcDNA:LD24607, CG14672, Dmel\CG14672, Dmel_CG14672, SPEC, SPEC2, Spec, dSPEC, dSPEC2
- Chromosome Location
-
Chr.3R: 5616644 - 5624125 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0044823
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169081.4 |
NP_730968.1 |
2 - 3 |
739 |
107 |
GenBank mRNA |
AY060373.1 |
- |
2 - 3 |
698 |
107 |
View Full Product Details