Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TrpRS
- Entrez Gene ID
-
45399
- Gene Name
- Tryptophanyl-tRNA synthetase
- Gene Aliases
-
85D-WRS, Aats-trp, CG9735, Dmel\CG9735, Dmel_CG9735, WRS, WRS-85D, dWARS, l(3)03559, l(3)03560, l(3)3559, wars
- Chromosome Location
-
Chr.3R: 9265027 - 9266904 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010803
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080087.3 |
NP_524826.1 |
1 - 2 |
311 |
71 |
GenBank mRNA |
BT044253.1 |
- |
1 - 2 |
355 |
71 |
View Full Product Details