Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cad87A
- Entrez Gene ID
-
41441
- Gene Name
- Cadherin 87A
- Gene Aliases
-
CG6977, CT21611, Cad87, DCad87A, Dmel\CG6977, Dmel_CG6977
- Chromosome Location
-
Chr.3R: 11893512 - 11949129 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037963
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169440.3 |
NP_731649.2 |
4 - 5 |
648 |
112 |
View Full Product Details