Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Lgr3
- Entrez Gene ID
-
43098
- Gene Name
- Leucine-rich repeat-containing G protein-coupled receptor 3
- Gene Aliases
-
CG31096, CG5042, CG5046, CT16193, DLGR3, Dmel\CG31096, Dmel_CG31096, anon-WO0170980.25, anon-WO0170980.26, dLGR3, lgr3
- Chromosome Location
-
Chr.3R: 25665809 - 25677214 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039354
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170236.2 |
NP_733115.1 |
8 - 9 |
1575 |
87 |
GenBank mRNA |
BT150256.1 |
- |
8 - 9 |
1575 |
87 |
View Full Product Details