Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crb
Entrez Gene ID
42896
Gene Name
crumbs
Gene Aliases
0509/20, 1384/04, CG6383, CRB, CT19912, Crb, Crbs, Crumbs, DmCrb, Dmel\CG6383, Dmel_CG6383, crumb, far, l(3)07207, l(3)S050920, l(3)S058104, l(3)j1B5
Chromosome Location
Chr.3R: 24295078 - 24314541 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259685
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043286.2 NP_001036751.1 13 - 14 6890 86
NM_001260355.2 NP_001247284.1 13 - 14 7082 86
NM_001275980.1 NP_001262909.1 14 - 15 6932 86
NM_079756.3 NP_524480.2 12 - 13 6761 86

View Full Product Details