Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Esp
Entrez Gene ID
42962
Gene Name
Epidermal stripes and patches
Gene Aliases
CG7005, Dmel\CG7005, Dmel_CG7005
Chromosome Location
Chr.3R: 24841935 - 24850412 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013953
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202353.2 NP_001189282.1 4 - 5 1744 76
NM_079766.4 NP_524490.2 3 - 4 1383 76
GenBank mRNA AY071258.1 - 3 - 4 1380 76

View Full Product Details