Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hr96
Entrez Gene ID
42993
Gene Name
Hormone receptor-like in 96
Gene Aliases
CG11783, DHR96, Dhr96, Dmel\CG11783, Dmel_CG11783, HR96, NR1J1, dHR96
Chromosome Location
Chr.3R: 25025235 - 25029351 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015240
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079769.3 NP_524493.1 3 - 4 1860 66
GenBank mRNA AY051486.1 - 3 - 4 1842 66
U36792.1 - 3 - 4 1818 66

View Full Product Details