Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gwl
- Entrez Gene ID
-
45969
- Gene Name
- greatwall
- Gene Aliases
-
1J, CG7719, CG7719-related, CT23435, Dmel\CG7719, Dmel_CG7719, EP0515, Gwl, Pk91C, Pk?2, anon-WO2004063362.73, scant
- Chromosome Location
-
Chr.3R: 18746370 - 18750360 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260399
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080121.4 |
NP_524860.2 |
4 - 5 |
1476 |
72 |
GenBank mRNA |
AY058648.1 |
- |
4 - 5 |
1448 |
72 |
View Full Product Details