The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260206.1 | NP_001247135.1 | 4 - 5 | 1634 | 89 |
NM_080257.4 | NP_524996.1 | 3 - 4 | 1667 | 89 | |
GenBank mRNA | AF227209.1 | - | 3 - 4 | 1651 | 89 |