Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mtSSB
- Entrez Gene ID
-
41968
- Gene Name
- mitochondrial single stranded DNA-binding protein
- Gene Aliases
-
0046/13, CG4337, Dmel\CG4337, Dmel_CG4337, DmmtSSB, SSB, SSB_DROME, c3a, dmtSSB, lopo
- Chromosome Location
-
Chr.3R: 16204194 - 16205250 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010438
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080496.4 |
NP_536744.2 |
2 - 3 |
130 |
58 |
GenBank mRNA |
BT016028.1 |
- |
2 - 3 |
119 |
58 |
View Full Product Details