Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr98d
- Entrez Gene ID
-
43309
- Gene Name
- Gustatory receptor 98d
- Gene Aliases
-
98B.1, CG12884, CG31061, Dmel\CG31061, Dmel_CG31061, GR98B.1, GR98d, Gr98A1, Gr98B1
- Chromosome Location
-
Chr.3R: 27546052 - 27547462 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046885
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170335.1 |
NP_733214.1 |
3 - 4 |
1112 |
147 |
View Full Product Details