Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cad99C
Entrez Gene ID
43528
Gene Name
Cadherin 99C
Gene Aliases
CA-10, CG15511, CG310034, CG31009, CG7805, CT35624, CT4930, Cad99c, DCad99C, Dm Cad99C, Dmel\CG31009, Dmel_CG31009, cad99C
Chromosome Location
Chr.3R: 29841657 - 29854699 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039709
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202382.1 NP_001189311.1 10 - 11 5125 57
NM_170435.2 NP_733314.1 11 - 12 5274 57

View Full Product Details