Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PH4alphaNE1
- Entrez Gene ID
-
43625
- Gene Name
- prolyl-4-hydroxylase-alpha NE1
- Gene Aliases
-
CG9728, Dmel\CG9728, Dmel_CG9728
- Chromosome Location
-
Chr.3R: 30496751 - 30500326 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039780
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170495.2 |
NP_733374.1 |
1 - 2 |
285 |
63 |
GenBank mRNA |
BT132956.1 |
- |
1 - 2 |
285 |
63 |
View Full Product Details