Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- pnt
- Entrez Gene ID
-
42757
- Gene Name
- pointed
- Gene Aliases
-
0123/09, 0608/07, 0998/12, 3520, CG17077, D-Ets-2, D-ets-2, DMPOINT1A, Dmel\CG17077, Dmel_CG17077, E(E2F)3D, EC3-2, EK3-2, ETS2, EY3-1, Ets, Ets1/2, Ets2, Ets58AB, Ets94F, PNT, PNT-P1, PNTP1, PNTP2, POINT, Pnt, Pnt-P1, Pnt2, PntP1, PntP2, Pointed, Pointed-P1, ets94F, l(3)07825, l(3)j1B7, l(3)s118306, png, pnt-P1, pnt-P2, pnt.P1, pntP2, pntegfr, pointed-RC, ptd
- Chromosome Location
-
Chr.3R: 23290231 - 23346167 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003118
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170651.4 |
NP_732857.1 |
1 - 2 |
652 |
91 |
GenBank mRNA |
BT133301.1 |
- |
2 - 3 |
610 |
91 |
View Full Product Details