Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mask
Entrez Gene ID
50070
Gene Name
multiple ankyrin repeats single KH domain
Gene Aliases
CG18671, CG31138, CG33106, CG6268, CG6313, Dmel\CG33106, Dmel_CG33106, EC3-6, MASK, Mask, cg6313
Chromosome Location
Chr.3R: 24230562 - 24250248 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0043884
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275975.1 NP_001262904.1 4 - 5 1786 76
NM_176556.2 NP_788733.1 4 - 5 1786 76
NM_176557.2 NP_788734.1 4 - 5 1786 76

View Full Product Details