Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- p53
- Entrez Gene ID
-
2768677
- Gene Name
- CG33336-PA
- Gene Aliases
-
CG10873, CG31325, CG33336, D-p53, DMP53, Dm-P53, DmP53, Dmel\CG33336, Dmel_CG33336, Dmp53, Dp53, P53, dmP53, dmp53, dp53, prac
- Chromosome Location
-
Chr.3R: 23049657 - 23054082 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039044
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206544.2 |
NP_996267.1 |
2 - 3 |
448 |
113 |
GenBank mRNA |
DQ191318.1 |
- |
2 - 3 |
388 |
113 |
View Full Product Details