Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Trc8
- Entrez Gene ID
-
43476
- Gene Name
- TRC8 ring finger protein
- Gene Aliases
-
CG2304, DTrc8, Dmel\CG2304, Dmel_CG2304, Dtrc8, TRC8, trc8
- Chromosome Location
-
Chr.3R: 29499025 - 29503797 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039668
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206580.2 |
NP_996303.1 |
4 - 5 |
2103 |
57 |
View Full Product Details