Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kaz1-ORFB
Entrez Gene ID
3772691
Gene Name
CG1220-PA
Gene Aliases
CG1220, CG33045, Dmel\CG1220, Dmel_CG1220, Kaz1, ORF B, ORF C, ORF D, ORF E
Chromosome Location
Chr.3L: 216625 - 218058 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0063923
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138169.5 NP_612013.3 2 - 3 358 150
NM_167809.4 NP_728495.1 4 - 5 939 150
NM_167810.3 NP_728496.2 3 - 4 926 150
GenBank mRNA AB017972.1 - 4 - 5 919 150
AB017973.1 - 3 - 4 926 150
AB017974.1 - 2 - 3 343 150
AY069042.1 - 2 - 3 339 150

View Full Product Details