Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sina
Entrez Gene ID
39884
Gene Name
seven in absentia
Gene Aliases
CG9949, Dmel\CG9949, Dmel_CG9949, SINA, Sina, anon-EST:fe2H7, sia
Chromosome Location
Chr.3L: 16853751 - 16858712 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003410
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300160.1 NP_001287089.1 3 - 3 1688 67
NM_057377.4 NP_476725.1 2 - 2 1755 67
NM_168689.2 NP_730206.1 2 - 2 1046 67
GenBank mRNA AY060358.1 - 2 - 2 1112 67
M38384.1 - 2 - 2 2761 67

View Full Product Details