Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rlb1
Entrez Gene ID
41139
Gene Name
CG8161-PA
Gene Aliases
CG8161, Dmel\CG8161, Dmel_CG8161, Ov30, Rlb
Chromosome Location
Chr.3R: 9513327 - 9514827 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014022
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057751.4 NP_477099.1 2 - 2 1400 91
GenBank mRNA AY119200.1 - 2 - 2 1372 91
X73220.1 - - 1381 91

View Full Product Details