Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Act57B
- Entrez Gene ID
-
37368
- Gene Name
- Actin 57B
- Gene Aliases
-
ACT3, Act57, Act57A, Act57b, Actin, ArpE, BEST:RE38067, C, CG10067, CG30294, Dmel\CG10067, Dmel_CG10067, M, T10, act 57A, act57A, act57B, act57b, actin, actin-57B, anon-EST:CL2c12, chr2R:16452771..16452966
- Chromosome Location
-
Chr.2R: 20944021 - 20946431 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000044
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079076.4 |
NP_523800.1 |
2 - 2 |
1542 |
66 |
GenBank mRNA |
BT099815.1 |
- |
2 - 2 |
1518 |
66 |
View Full Product Details