Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TwdlH
Entrez Gene ID
43208
Gene Name
TweedleH
Gene Aliases
CG14241, CG31080, Dmel\CG31080, Dmel_CG31080
Chromosome Location
Chr.3R: 26633732 - 26634714 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051080
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143233.3 NP_651490.1 2 - 2 849 84
GenBank mRNA AY070579.1 - 2 - 2 847 84

View Full Product Details