Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kermit
Entrez Gene ID
46027
Gene Name
CG11546-PA
Gene Aliases
BEST:LD04728, CG11546, Dmel\CG11546, Dmel_CG11546, LD04728, dGIPC, dKermit, l(2)02045
Chromosome Location
Chr.2R: 8146918 - 8156654 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010504
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299258.1 NP_001286187.1 1 - 1 1439 146
NM_143771.4 NP_652028.1 2 - 2 1856 146
NM_165590.3 NP_724649.1 2 - 2 1497 146
NM_165591.3 NP_724650.1 2 - 2 1500 146
NM_165592.3 NP_724651.1 2 - 2 1384 146
GenBank mRNA AY060451.1 - 2 - 2 1777 146

View Full Product Details