Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BomS2
- Entrez Gene ID
-
49802
- Gene Name
- Bomanin Short 2
- Gene Aliases
-
BcDNA:RH08291, Bom2, CG18106, DIM 2, DIM-2, DIM2, Dmel\CG18106, Dmel_CG18106, IM2, Im2, anon-WO0140519.2
- Chromosome Location
-
Chr.2R: 18386602 - 18387030 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0025583
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166277.3 |
NP_725776.1 |
2 - 2 |
257 |
88 |
GenBank mRNA |
AY071680.1 |
- |
2 - 2 |
257 |
88 |
View Full Product Details