Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hb
Entrez Gene ID
41032
Gene Name
hunchback
Gene Aliases
CG9786, Dmel\CG9786, Dmel_CG9786, HB, Hb, Hunchback, R-pbx, Rg-bx, Rg-pbx, hbHLH, l(3)85Ah
Chromosome Location
Chr.3R: 8690980 - 8697822 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001180
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169233.2 NP_731267.1 2 - 2 2043 80
NM_169234.2 NP_731268.1 2 - 2 2395 80
GenBank mRNA BT024234.1 - 2 - 2 2365 80

View Full Product Details