Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rab9E
- Entrez Gene ID
-
318148
- Gene Name
- Rab at 9E
- Gene Aliases
-
CG32673, Dmel\CG32673, Dmel_CG32673, anon-AE003451.2
- Chromosome Location
-
Chr.X: 10660955 - 10661843 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052673
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167238.3 |
NP_727438.1 |
1 - 1 |
256 |
152 |
View Full Product Details