Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Gpb5
Entrez Gene ID
3355097
Gene Name
Glycoprotein hormone beta 5
Gene Aliases
CG40041, Dmbeta, Dmel\CG40041, Dmel_CG40041, GPB5, beta-glycoprotein-hormone-related, gpb5
Chromosome Location
Chr.2L: 23311700 - 23312464 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0063368
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015386.3 NP_001015386.3 2 - 2 297 82
NM_001110864.2 NP_001104334.1 1 - 1 334 82
NM_001110865.3 NP_001104335.1 2 - 2 257 82
GenBank mRNA AF403389.1 - 1 - 1 227 82
AJ440748.1 - 1 - 1 250 82
BT023078.1 - 2 - 2 272 82

View Full Product Details