Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
kl-2
Entrez Gene ID
3355181
Gene Name
male fertility factor kl2
Gene Aliases
CG17866, CG40438, DHC3A, Dmel\CG17866, Dmel_CG17866, Lms2, kl2
Chromosome Location
Chr.Y: 573088 - 664173 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001313
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015505.5 NP_001015505.5 11 - 12 12832 121

View Full Product Details