Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hk
Entrez Gene ID
31955
Gene Name
Hyperkinetic
Gene Aliases
CG17779, CG2287, CG32688, CG34319, CG43388, Dm_X:51771, Dmel\CG43388, Dmel_CG43388, hk, lincRNA.975
Chromosome Location
Chr.X: 10233347 - 10263485 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263220
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258681.2 NP_001245610.1 1 - 2 554 78

View Full Product Details