Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dexi
Entrez Gene ID
445307
Gene Name
Dexi homolog (mouse)
Gene Aliases
im:7136011, zgc:101082
Chromosome Location
Chr.3: 26905024 - 26906351 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003764.3 NP_001003764.1 1 - 2 809 81
GenBank mRNA BC079513.1 - 1 - 2 781 81

View Full Product Details