Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eef2kmt
Entrez Gene ID
450038
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
fam86a, zgc:101751
Chromosome Location
Chr.3: 28869610 - 28872929 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006058.1 NP_001006058.1 5 - 6 561 70
GenBank mRNA BC083266.1 - 5 - 6 561 70

View Full Product Details