Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pop5
Entrez Gene ID
541421
Gene Name
POP5 homolog, ribonuclease P/MRP subunit
Gene Aliases
fc69d03, wu:fc69d03, wu:fy24c05, zgc:110616
Chromosome Location
Chr.8: 41274208 - 41279294 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013548.1 NP_001013566.1 3 - 4 350 90
GenBank mRNA BC091467.1 - 3 - 4 350 90

View Full Product Details