Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
il10ra
Entrez Gene ID
777651
Gene Name
interleukin 10 receptor, alpha
Gene Aliases
crfb7
Chromosome Location
Chr.15: 12377994 - 12388434 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077625.1 NP_001071093.1 2 - 3 301 176
GenBank mRNA BC142763.1 - 2 - 3 408 176
EF014957.1 - 2 - 3 301 176

View Full Product Details