Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il10ra
Entrez Gene ID
777651
Gene Name
interleukin 10 receptor, alpha
Gene Aliases
crfb7
Chromosome Location
Chr.15: 12377994 - 12388434 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077625.1 NP_001071093.1 6 - 7 888 60
GenBank mRNA EF014957.1 - 6 - 7 888 60

View Full Product Details