Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ryr1b
Entrez Gene ID
570245
Gene Name
ryanodine receptor 1b (skeletal)
Gene Aliases
im:7137738, ryrab, si:dkey-250i19.1
Chromosome Location
Chr.18: 35489483 - 35736799 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102571.1 NP_001096041.1 74 - 75 10738 84
XM_017351957.2 XP_017207446.1 75 - 76 84
XM_017351958.2 XP_017207447.1 74 - 75 84
XM_017351959.2 XP_017207448.1 74 - 75 84
XM_017351960.2 XP_017207449.1 75 - 76 84
XM_017351961.2 XP_017207450.1 74 - 75 84
XM_017351962.2 XP_017207451.1 74 - 75 84
XM_017351963.2 XP_017207452.1 73 - 74 84
XM_021467693.1 XP_021323368.1 75 - 76 84
XM_021467694.1 XP_021323369.1 75 - 76 84
XM_021467695.1 XP_021323370.1 74 - 75 84
XM_021467696.1 XP_021323371.1 75 - 76 84
GenBank mRNA AB247454.1 - 74 - 75 10738 84

View Full Product Details