Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cahz
Entrez Gene ID
30331
Gene Name
carbonic anhydrase
Gene Aliases
CAH-Z, cah, cb588, id:ibd5016, wu:cegs2190, wu:fj65c09
Chromosome Location
Chr.2: 29477383 - 29485105 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131110.1 NP_571185.1 2 - 3 289 115
GenBank mRNA BC065611.1 - 2 - 3 285 115
U55177.1 - 2 - 3 289 115

View Full Product Details