Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ddx39b
Entrez Gene ID
393917
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B
Gene Aliases
Bat1a, bat1, zgc:63773
Chromosome Location
Chr.19: 27339848 - 27357682 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200943.3 NP_957237.1 2 - 3 320 65
GenBank mRNA BC055240.1 - 2 - 3 291 65
EH447839.1 - 2 - 3 322 65
EH457748.1 - 2 - 3 324 65

View Full Product Details