Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptbp1a
Entrez Gene ID
323490
Gene Name
polypyrimidine tract binding protein 1a
Gene Aliases
ptbp1, wu:fb99f12, zgc:110689
Chromosome Location
Chr.2: 26288287 - 26300617 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020477.1 NP_001018313.1 5 - 6 352 97
XM_021478816.1 XP_021334491.1 5 - 6 419 97
GenBank mRNA BC095372.1 - 5 - 6 352 97

View Full Product Details