Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec62
Entrez Gene ID
563660
Gene Name
SEC62 homolog, preprotein translocation factor
Gene Aliases
fb57b06, tloc1, wu:fb57b06, zgc:114073
Chromosome Location
Chr.2: 37245393 - 37264229 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025530.1 NP_001020701.1 6 - 7 679 118
GenBank mRNA BC097135.1 - 6 - 7 679 118

View Full Product Details