Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trmt2a
Entrez Gene ID
334936
Gene Name
tRNA methyltransferase 2 homolog A
Gene Aliases
fa10b06, htf9c, wu:fa10b06, zgc:55519
Chromosome Location
Chr.5: 228296 - 239848 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199929.1 NP_956223.1 2 - 3 753 112
GenBank mRNA BC045367.1 - 2 - 3 753 112

View Full Product Details