Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srm
Entrez Gene ID
394009
Gene Name
spermidine synthase
Gene Aliases
fa18b03, id:ibd1177, wu:fa18b03, zgc:63600
Chromosome Location
Chr.23: 28718859 - 28727106 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201034.2 NP_957328.2 1 - 2 265 72
GenBank mRNA BC055159.1 - 1 - 2 210 72
CT621066.2 - 1 - 2 124 72

View Full Product Details