Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klf12a
Entrez Gene ID
798629
Gene Name
Kruppel-like factor 12a
Gene Aliases
fb02h07, klf12, wu:fb02h07, zgc:77286
Chromosome Location
Chr.1: 108725 - 163840 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207062.1 NP_996945.1 4 - 5 858 73
XM_021475848.1 XP_021331523.1 5 - 6 1133 73
GenBank mRNA BC066429.1 - 4 - 5 858 73

View Full Product Details