Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prpf38b
Entrez Gene ID
406810
Gene Name
pre-mRNA processing factor 38B
Gene Aliases
wu:fc43b05, zgc:63496
Chromosome Location
Chr.2: 45655294 - 45663929 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213489.1 NP_998654.1 2 - 3 490 117
XM_009298770.3 XP_009297045.1 2 - 3 511 117
GenBank mRNA BC061448.1 - 2 - 3 490 117

View Full Product Details