Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aebp2
Entrez Gene ID
406815
Gene Name
AE binding protein 2
Gene Aliases
wu:fk42f11, wu:fk52h12, zgc:63755
Chromosome Location
Chr.4: 2927270 - 2945327 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213494.2 NP_998659.2 1 - 2 546 74
GenBank mRNA BC055234.1 - 1 - 2 143 74

View Full Product Details