Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
b4galt7
Entrez Gene ID
445022
Gene Name
xylosylprotein beta 1,4-galactosyltransferase, polypeptide 7 (galactosyltransferase I)
Gene Aliases
zgc:110571
Chromosome Location
Chr.14: 26380451 - 26392198 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003417.1 NP_001003417.1 2 - 3 904 131
XM_021481002.1 XP_021336677.1 3 - 4 990 131
GenBank mRNA AY648740.1 - 2 - 3 904 131
BC092934.1 - 2 - 3 842 131
BC164341.1 - 2 - 3 419 131

View Full Product Details