Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
uxt
Entrez Gene ID
447933
Gene Name
ubiquitously-expressed, prefoldin-like chaperone
Gene Aliases
zgc:101894
Chromosome Location
Chr.8: 8699085 - 8704430 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004671.1 NP_001004671.1 3 - 4 448 81
GenBank mRNA BC081425.1 - 3 - 4 448 81

View Full Product Details