Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nova1
Entrez Gene ID
555590
Gene Name
NOVA alternative splicing regulator 1
Gene Aliases
si:ch211-222c22.9
Chromosome Location
Chr.5: 36611204 - 36632068 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044789.1 NP_001038254.1 1 - 2 161 86
GenBank mRNA BC171495.1 - 1 - 2 237 86
BC171499.1 - 1 - 2 237 86

View Full Product Details