Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nova1
Entrez Gene ID
555590
Gene Name
NOVA alternative splicing regulator 1
Gene Aliases
si:ch211-222c22.9
Chromosome Location
Chr.5: 36611204 - 36632068 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044789.1 NP_001038254.1 3 - 4 474 75
XM_005165318.4 XP_005165375.1 3 - 4 833 75
GenBank mRNA BC171495.1 - 3 - 4 550 75
BC171499.1 - 3 - 4 550 75

View Full Product Details